(3-Bromo-2-fluoro-5,6-dimethoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

53445-500MG 53445-1G 53445-5G 53445-25G
77329-016EA 140.08 USD
77329-016 77344-750 77365-304 77387-344
(3-Bromo-2-fluoro-5,6-dimethoxyphenyl)(methyl)sulfane ≥97%
(3-Bromo-2-fluoro-5,6-dimethoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₀BrFO₂S
Storage Temperature: Ambient
MDL Number: MFCD34178697
CAS Number: 2643368-59-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR