(2-Bromo-3-methoxyphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

53437-1G 53437-5G 53437-25G
77341-694EA 100.94 USD
77341-694 77365-300 77377-034
(2-Bromo-3-methoxyphenyl)boronic acid ≥97%
(2-Bromo-3-methoxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₈BBrO₃
MW: 230.85 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28040469
CAS Number: 849630-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR