(3-Ethyl-5-iodophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

53428-500MG 53428-1G 53428-5G
77338-916EA 530.45 USD
77338-916 77352-800 77377-032
(3-Ethyl-5-iodophenyl)methanol ≥97%
(3-Ethyl-5-iodophenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₁IO
MW: 262.088 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530218
CAS Number: 2734776-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR