1-(5-Bromo-2-methylphenyl)piperidine ≥97%

Supplier: AOBChem USA

Warning

53410-1G 53410-5G 53410-25G
77341-232EA 77.25 USD
77341-232 77365-296 77377-024
1-(5-Bromo-2-methylphenyl)piperidine ≥97%
1-(5-Bromo-2-methylphenyl)piperidine

Precaución: for Research Use Only

Formula: C₁₂H₁₆BrN
MW: 254.166 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24133831
CAS Number: 1500843-29-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR