1-(4-Bromo-5-fluoro-2-methylphenyl)piperidine ≥97%

Supplier: AOBChem USA

Warning

53408-500MG 53408-1G 53408-5G 53408-25G
77329-010EA 214.24 USD
77329-010 77352-794 77365-294 77387-340
1-(4-Bromo-5-fluoro-2-methylphenyl)piperidine ≥97%
1-(4-Bromo-5-fluoro-2-methylphenyl)piperidine

Precaución: for Research Use Only

Formula: C₁₂H₁₅BrFN
Storage Temperature: Ambient
MDL Number: MFCD34530217
CAS Number: 1771110-70-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR