4-(tert-Butyl)-2-iodo-1-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

53398-500MG 53398-1G 53398-5G 53398-25G
77329-008EA 166.86 USD
77329-008 77346-210 77365-290 77387-338
4-(tert-Butyl)-2-iodo-1-methylbenzene ≥95%
4-(tert-Butyl)-2-iodo-1-methylbenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₅I
MW: 274.141 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29096154
CAS Number: 70728-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR