3-Bromo-4-ethoxy-2-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

53392-500MG 53392-1G 53392-5G
77335-960EA 370.8 USD
77335-960 77352-792 77377-020
3-Bromo-4-ethoxy-2-fluorobenzoic acid ≥97%
3-Bromo-4-ethoxy-2-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₉H₈BrFO₃
Storage Temperature: Ambient
MDL Number: MFCD34757023
CAS Number: 2384034-72-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR