(2-Ethoxy-6-fluorophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

53386-1G 53386-5G 53386-25G
77341-924EA 108.15 USD
77341-924 77365-288 77377-016
(2-Ethoxy-6-fluorophenyl)(methyl)sulfane ≥97%
(2-Ethoxy-6-fluorophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₁FOS
MW: 186.246 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564234
CAS Number: 862401-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR