1-Ethoxy-3-fluoro-2-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

53385-500MG 53385-1G 53385-5G 53385-25G
77329-004EA 226.6 USD
77329-004 77352-788 77365-286 77387-336
1-Ethoxy-3-fluoro-2-iodobenzene ≥97%
1-Ethoxy-3-fluoro-2-iodobenzene

Precaución: for Research Use Only

Formula: C₈H₈FIO
Storage Temperature: Ambient
MDL Number: MFCD34564233
CAS Number: 1849329-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR