4-Acetyl-2-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

53381-500MG 53381-1G 53381-5G 53381-25G
77329-000EA 206 USD
77329-000 77352-784 77365-282 77387-332
4-Acetyl-2-methylbenzaldehyde ≥97%
4-Acetyl-2-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD31696746
CAS Number: 1433821-65-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR