(5-Bromopyridin-3-yl)(thiomorpholino)methanone ≥95%

Supplier: AOBChem USA

Warning

53377-500MG 53377-1G 53377-5G 53377-25G
77328-998EA 85.49 USD
77328-998 77342-218 77365-280 77377-014
(5-Bromopyridin-3-yl)(thiomorpholino)methanone ≥95%
(5-Bromopyridin-3-yl)(thiomorpholino)methanone

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrN₂OS
Storage Temperature: Ambient
MDL Number: MFCD13575556
CAS Number: 1197957-37-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR