(5-Bromopyridin-3-yl)(4-methylpiperazin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

53376-500MG 53376-1G 53376-5G 53376-25G
77328-996EA 212.18 USD
77328-996 77352-782 77365-278 77387-330
(5-Bromopyridin-3-yl)(4-methylpiperazin-1-yl)methanone ≥97%
(5-Bromopyridin-3-yl)(4-methylpiperazin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₁H₁₄BrN₃O
Storage Temperature: Ambient
MDL Number: MFCD00448863
CAS Number: 342013-83-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR