5-Bromo-N-isobutylnicotinamide ≥97%

Supplier: AOBChem USA

Warning

53374-1G 53374-5G 53374-25G
77340-958EA 64.89 USD
77340-958 77365-276 77377-012
5-Bromo-N-isobutylnicotinamide ≥97%
5-Bromo-N-isobutylnicotinamide

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD10653572
CAS Number: 1088170-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR