(3-Fluoro-4,5-dimethoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

53373-500MG 53373-1G 53373-5G
77337-532EA 417.15 USD
77337-532 77352-780 77377-010
(3-Fluoro-4,5-dimethoxyphenyl)methanol ≥97%
(3-Fluoro-4,5-dimethoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₁FO₃
MW: 186.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12756552
CAS Number: 99423-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR