Ethyl 3-fluoro-4,5-dimethoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

53372-500MG 53372-1G 53372-5G
77338-604EA 503.67 USD
77338-604 77352-778 77377-008
Ethyl 3-fluoro-4,5-dimethoxybenzoate ≥97%
Ethyl 3-fluoro-4,5-dimethoxybenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₄
MW: 228.217 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12911444
CAS Number: 1260765-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR