2-(3-Fluoro-4,5-dimethoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

53369-500MG 53369-1G 53369-5G
77339-040EA 546.93 USD
77339-040 77352-774 77377-004
2-(3-Fluoro-4,5-dimethoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(3-Fluoro-4,5-dimethoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₄H₂₀BFO₄
MW: 282.116 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757017
CAS Number: 2477881-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR