(3-Fluoro-4,5-dimethoxyphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

53368-500MG 53368-1G 53368-5G
77338-486EA 490.28 USD
77338-486 77352-772 77377-002
(3-Fluoro-4,5-dimethoxyphenyl)boronic acid ≥97%
(3-Fluoro-4,5-dimethoxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BFO₄
MW: 199.972 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28793137
CAS Number: 1840935-37-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR