3-Chloro-2-fluoro-4-formylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

53361-500MG 53361-1G 53361-5G
77328-994EA 336.81 USD
77328-994 77352-770 77377-000
3-Chloro-2-fluoro-4-formylbenzoic acid ≥97%
3-Chloro-2-fluoro-4-formylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₄ClFO₃
MW: 202.567 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757013
CAS Number: 2387369-95-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR