2-Chloro-3-fluoro-4-iodobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

53360-500MG 53360-1G
77340-568EA 2111.5 USD
77340-568 77352-768
2-Chloro-3-fluoro-4-iodobenzaldehyde ≥97%
2-Chloro-3-fluoro-4-iodobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃ClFIO
MW: 284.454 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32875976
CAS Number: 2385079-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR