(4-(Benzyloxy)-3-bromo-2-fluorophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

53354-500MG 53354-1G 53354-5G
77339-952EA 692.16 USD
77339-952 77352-766 77376-998
(4-(Benzyloxy)-3-bromo-2-fluorophenyl)methanol ≥97%
(4-(Benzyloxy)-3-bromo-2-fluorophenyl)methanol

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrFO₂
MW: 311.146 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757009
CAS Number: 2807463-96-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR