3-Chloro-5-cyanobenzoic acid ≥95%

Supplier: AOBChem USA

Warning

53329-500MG 53329-1G 53329-5G 53329-25G
77328-984EA 127.72 USD
77328-984 77344-210 77365-266 77376-996
3-Chloro-5-cyanobenzoic acid ≥95%
3-Chloro-5-cyanobenzoic acid

Precaución: for Research Use Only

Formula: C₈H₄ClNO₂
MW: 181.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06797228
CAS Number: 327056-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR