(4-Iodo-2-(trifluoromethyl)phenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

53317-500MG 53317-1G 53317-5G 53317-25G
77328-982EA 114.33 USD
77328-982 77343-360 77365-264 77376-994
(4-Iodo-2-(trifluoromethyl)phenyl)(methyl)sulfane ≥97%
(4-Iodo-2-(trifluoromethyl)phenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₆F₃IS
MW: 318.098 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34178694
CAS Number: 2643368-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR