5-Chloro-1-fluoro-3-iodo-2-isopropoxybenzene ≥97%

Supplier: AOBChem USA

Warning

53315-500MG 53315-1G 53315-5G 53315-25G
77328-978EA 234.84 USD
77328-978 77352-754 77365-260 77387-318
5-Chloro-1-fluoro-3-iodo-2-isopropoxybenzene ≥97%
5-Chloro-1-fluoro-3-iodo-2-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₉H₉ClFIO
MW: 314.523 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530214
CAS Number: 2734778-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR