5-Chloro-2-(difluoromethoxy)-1-fluoro-3-iodobenzene ≥97%
Supplier: AOBChem USA
|
Warning
|
53314-500MG
53314-1G
53314-25G
53314-5G
77328-976EA
299.73
USD
77328-976
77352-752
77387-316
77376-992
5-Chloro-2-(difluoromethoxy)-1-fluoro-3-iodobenzene ≥97%
5-Chloro-2-(difluoromethoxy)-1-fluoro-3-iodobenzene
Precaución: for Research Use Only
Formula:
C₇H₃ClF₃IO MW: 322.451 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34530213 CAS Number: 2384792-39-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...