1-Bromo-5-chloro-2-(difluoromethoxy)-3-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

53303-500MG 53303-1G 53303-5G
77336-688EA 391.4 USD
77336-688 77352-748 77376-988
1-Bromo-5-chloro-2-(difluoromethoxy)-3-fluorobenzene ≥97%
1-Bromo-5-chloro-2-(difluoromethoxy)-3-fluorobenzene

Precaución: for Research Use Only

Formula: C₇H₃BrClF₃O
Storage Temperature: Ambient
MDL Number: MFCD22580814
CAS Number: 1417567-82-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR