Ethyl 4-ethoxy-3-fluoro-2-methylbenzoate ≥95%

Supplier: AOBChem USA

Warning

53299-500MG 53299-1G 53299-5G
77338-648EA 506.76 USD
77338-648 77352-742 77376-980
Ethyl 4-ethoxy-3-fluoro-2-methylbenzoate ≥95%
Ethyl 4-ethoxy-3-fluoro-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₅FO₃
MW: 226.244 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34178690
CAS Number: 2385741-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR