Methyl 4-ethoxy-3-fluoro-2-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

53298-500MG 53298-1G 53298-5G
77338-250EA 470.71 USD
77338-250 77352-740 77376-978
Methyl 4-ethoxy-3-fluoro-2-methylbenzoate ≥97%
Methyl 4-ethoxy-3-fluoro-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₃
Storage Temperature: Ambient
MDL Number: MFCD34178689
CAS Number: 2383988-42-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR