4-Ethoxy-3-fluoro-2-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

53297-500MG 53297-1G 53297-5G
77336-812EA 394.49 USD
77336-812 77352-738 77376-976
4-Ethoxy-3-fluoro-2-methylbenzoic acid ≥97%
4-Ethoxy-3-fluoro-2-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₃
MW: 198.191 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34178688
CAS Number: 2384775-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR