4-Ethoxy-3-fluoro-2-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

53295-500MG 53295-1G 53295-5G
77336-742EA 393.46 USD
77336-742 77352-736 77376-974
4-Ethoxy-3-fluoro-2-methylbenzaldehyde ≥97%
4-Ethoxy-3-fluoro-2-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₂
Storage Temperature: Ambient
MDL Number: MFCD34178687
CAS Number: 2386887-79-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR