(4-Ethoxy-3-fluoro-2-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

53294-500MG 53294-1G 53294-5G
77338-186EA 468.65 USD
77338-186 77352-734 77376-972
(4-Ethoxy-3-fluoro-2-methylphenyl)(methyl)sulfane ≥97%
(4-Ethoxy-3-fluoro-2-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₀H₁₃FOS
Storage Temperature: Ambient
MDL Number: MFCD34178686
CAS Number: 2643367-89-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR