4-(4-Bromophenyl)-2-chloropyridine ≥97%

Supplier: AOBChem USA

Warning

53273-500MG 53273-1G 53273-25G 53273-5G
77328-968EA 305.91 USD
77328-968 77352-730 77387-310 77376-968
4-(4-Bromophenyl)-2-chloropyridine ≥97%
4-(4-Bromophenyl)-2-chloropyridine

Precaución: for Research Use Only

Formula: C₁₁H₇BrClN
MW: 268.537 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28334954
CAS Number: 1449123-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR