2-Chloro-4-(m-tolyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

53272-500MG 53272-1G 53272-5G
77328-966EA 319.3 USD
77328-966 77352-728 77376-966
2-Chloro-4-(m-tolyl)pyridine ≥97%
2-Chloro-4-(m-tolyl)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₀ClN
MW: 203.667 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26640861
CAS Number: 1512550-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR