4-(4-Bromo-2,3-dimethylphenyl)-2-chloropyridine ≥97%

Supplier: AOBChem USA

Warning

53271-500MG 53271-1G 53271-5G 53271-25G
77328-964EA 226.6 USD
77328-964 77352-726 77365-248 77387-308
4-(4-Bromo-2,3-dimethylphenyl)-2-chloropyridine ≥97%
4-(4-Bromo-2,3-dimethylphenyl)-2-chloropyridine

Precaución: for Research Use Only

Formula: C₁₃H₁₁BrClN
MW: 296.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34178684
CAS Number: 2643367-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR