4-(4-Bromo-2,5-dimethylphenyl)-2-chloropyridine ≥95%

Supplier: AOBChem USA

Warning

53270-500MG 53270-1G 53270-5G 53270-25G
77328-962EA 226.6 USD
77328-962 77352-724 77365-246 77387-306
4-(4-Bromo-2,5-dimethylphenyl)-2-chloropyridine ≥95%
4-(4-Bromo-2,5-dimethylphenyl)-2-chloropyridine

Precaución: for Research Use Only

Formula: C₁₃H₁₁BrClN
MW: 296.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34178683
CAS Number: 2643367-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR