2-Chloro-4-(3,4-dimethylphenyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

53265-500MG 53265-1G 53265-5G
77328-958EA 346.08 USD
77328-958 77352-720 77376-964
2-Chloro-4-(3,4-dimethylphenyl)pyridine ≥97%
2-Chloro-4-(3,4-dimethylphenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₃H₁₂ClN
MW: 217.694 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26643967
CAS Number: 1505800-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR