2-Chloro-4-(4-methoxyphenyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

53264-500MG 53264-1G 53264-5G
77328-956EA 334.75 USD
77328-956 77352-718 77376-962
2-Chloro-4-(4-methoxyphenyl)pyridine ≥97%
2-Chloro-4-(4-methoxyphenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₀ClNO
MW: 219.667 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09907964
CAS Number: 914303-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR