4-(4-(Benzyloxy)-3-methylphenyl)-2-chloropyridine ≥95%

Supplier: AOBChem USA

Warning

53262-500MG 53262-1G 53262-5G
77336-086EA 375.95 USD
77336-086 77352-714 77376-958
4-(4-(Benzyloxy)-3-methylphenyl)-2-chloropyridine ≥95%
4-(4-(Benzyloxy)-3-methylphenyl)-2-chloropyridine

Precaución: for Research Use Only

Formula: C₁₉H₁₆ClNO
MW: 309.789 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530205
CAS Number: 2734772-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR