4-(4-(Benzyloxy)-3-fluorophenyl)-2-chloropyridine ≥97%

Supplier: AOBChem USA

Warning

53261-500MG 53261-1G 53261-5G
77335-472EA 355.35 USD
77335-472 77352-712 77376-956
4-(4-(Benzyloxy)-3-fluorophenyl)-2-chloropyridine ≥97%
4-(4-(Benzyloxy)-3-fluorophenyl)-2-chloropyridine

Precaución: for Research Use Only

Formula: C₁₈H₁₃ClFNO
MW: 313.753 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530204
CAS Number: 2734775-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR