2-Chloro-4-(3-Chloro-4-ethoxyphenyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

53259-500MG 53259-1G 53259-5G
77338-838EA 524.27 USD
77338-838 77352-708 77376-952
2-Chloro-4-(3-Chloro-4-ethoxyphenyl)pyridine ≥97%
2-Chloro-4-(3-Chloro-4-ethoxyphenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₃H₁₁Cl₂NO
Storage Temperature: Ambient
MDL Number: MFCD34530202
CAS Number: 2734775-89-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR