2-Chloro-4-(2,4-dimethoxyphenyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

53257-500MG 53257-1G 53257-5G
77336-418EA 387.28 USD
77336-418 77352-704 77376-948
2-Chloro-4-(2,4-dimethoxyphenyl)pyridine ≥97%
2-Chloro-4-(2,4-dimethoxyphenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₃H₁₂ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD34530200
CAS Number: 2734776-95-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR