2-Chloro-4-(4-Chloro-2-fluorophenyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

53247-500MG 53247-1G 53247-5G 53247-25G
77328-952EA 122.57 USD
77328-952 77343-998 77365-242 77376-944
2-Chloro-4-(4-Chloro-2-fluorophenyl)pyridine ≥95%
2-Chloro-4-(4-Chloro-2-fluorophenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₁H₆Cl₂FN
Storage Temperature: Ambient
MDL Number: MFCD34564231
CAS Number: 2734778-49-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR