2-Chloro-4-(2-fluoro-4-methylphenyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

53246-500MG 53246-1G 53246-5G
77337-152EA 408.91 USD
77337-152 77352-700 77376-942
2-Chloro-4-(2-fluoro-4-methylphenyl)pyridine ≥97%
2-Chloro-4-(2-fluoro-4-methylphenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₉ClFN
MW: 221.658 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530196
CAS Number: 2734779-62-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR