2-Chloro-4-(3-Chloro-4-methylphenyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

53245-500MG 53245-1G 53245-5G
77340-238EA 854.9 USD
77340-238 77352-698 77376-940
2-Chloro-4-(3-Chloro-4-methylphenyl)pyridine ≥97%
2-Chloro-4-(3-Chloro-4-methylphenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₉Cl₂N
MW: 238.113 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26651989
CAS Number: 1507845-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR