4-(4-Bromo-2-methylphenyl)-2-chloropyridine ≥97%

Supplier: AOBChem USA

Warning

53244-500MG 53244-1G 53244-5G
77336-084EA 375.95 USD
77336-084 77352-696 77376-938
4-(4-Bromo-2-methylphenyl)-2-chloropyridine ≥97%
4-(4-Bromo-2-methylphenyl)-2-chloropyridine

Precaución: for Research Use Only

Formula: C₁₂H₉BrClN
MW: 282.564 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26659534
CAS Number: 1541963-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR