4-Formyl-3-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

53223-500MG 53223-1G 53223-5G
77339-602EA 603.58 USD
77339-602 77352-694 77376-934
4-Formyl-3-(trifluoromethyl)benzoic acid ≥97%
4-Formyl-3-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₅F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD28357161
CAS Number: 1289083-41-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR