(4-Bromo-2-fluoro-3-methoxyphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

53221-1G 53221-5G 53221-25G
77341-426EA 85.49 USD
77341-426 77365-236 77376-932
(4-Bromo-2-fluoro-3-methoxyphenyl)(methyl)sulfane ≥95%
(4-Bromo-2-fluoro-3-methoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈BrFOS
MW: 251.116 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28132069
CAS Number: 1824272-66-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR