2-Fluoro-3,5-dimethylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

53218-500MG 53218-1G 53218-5G
77336-686EA 391.4 USD
77336-686 77352-690 77376-928
2-Fluoro-3,5-dimethylbenzoic acid ≥97%
2-Fluoro-3,5-dimethylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₂
MW: 168.165 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23712058
CAS Number: 1427083-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR