1-(2,3-Difluoro-4-iodophenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

53215-500MG 53215-1G 53215-5G 53215-25G
77328-946EA 100.94 USD
77328-946 77342-930 77365-234 77376-926
1-(2,3-Difluoro-4-iodophenyl)ethanol ≥97%
1-(2,3-Difluoro-4-iodophenyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₇F₂IO
MW: 284.042 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756991
CAS Number: 2755722-90-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR