Methyl 4-chloro-2-isopropoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

53206-500MG 53206-1G 53206-5G
77339-726EA 620.06 USD
77339-726 77352-684 77376-920
Methyl 4-chloro-2-isopropoxybenzoate ≥97%
Methyl 4-chloro-2-isopropoxybenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃ClO₃
MW: 228.672 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756988
CAS Number: 2755717-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR