4-Chloro-2-isopropoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

53205-500MG 53205-1G 53205-5G
77328-944EA 345.05 USD
77328-944 77352-682 77376-918
4-Chloro-2-isopropoxybenzoic acid ≥97%
4-Chloro-2-isopropoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₃
Storage Temperature: Ambient
MDL Number: MFCD14652941
CAS Number: 689161-75-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR